Learn More
Saint Gobain Performance Plastics Tube Tygon 1/8 x 1/4 100FT/PK
Supplier:  Saint Gobain Performance Plastics ACF02007
Encompass_Preferred
Saint-Gobain Tygon S3 E-3603 Flexible Tubing 100 ft. Diameter Inner 1/8 in. 3.2 mm Diameter Outer 1/4 in. 6.35 mm Thickness 1/16 in. 1.6 mm Overall Length 100 ft. 30.5 m
Catalog No. 50-206-8973
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Explore available promotions