Learn More
Saint Gobain Performance Plastics Tube Tygon 1/4 x 3/8 500FT/PK
Supplier: Saint Gobain Performance Plastics ACF04017
Saint-Gobain Tygon E-3603 Flexible Tubing Diameter Inner 6.4 mm Diameter Outer 9.5 mm Overall Length 152 m Thickness 1.6 mm