Learn More
Saint Gobain Performance Plastics TUB SURG 1/2 x 5/8 50FT-15M/PK
Supplier:  Saint Gobain Performance Plastics ADF00036
Encompass_Preferred
Saint-Gobain Tygon ND 100-65 Medical Tubings Diameter Inner 1/2 in. Diameter Outer 5/8 in. Thickness 1/16 in. Non-DEHP Overall Length 50 ft. 15.2 m Clear
Catalog No. 50-206-8993
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product