Learn More
Saint Gobain Performance Plastics TUB S-50-HL 3/32 x 5/32 50FT/PK
Supplier: Saint Gobain Performance Plastics ADF00004
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Saint-Gobain Tygon ND 100-65 Medical Tubings Diameter Inner 3/32 in. Diameter Outer 5/32 in. Thickness 1/32 in. Overall Length 50 ft. 15.2 m Clear Must order minimum quantity of 2.