Learn More
Saint Gobain Performance Plastics TUB S-50-HL 1/16 x 1/8 100FT/PK
Supplier: Saint Gobain Performance Plastics ADF02002
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Saint-Gobain Tygon ND 100-65 Medical Tubing Diameter Inner 1/16 in. 1.6 mm Diameter Outer 1/8 in. 3.2 mm Overall Length 100 ft. 30.4 m Thickness 1/32 in. Non-DEHP Must order minimum quantity of 2.