Learn More
Pfanstiehl Inc Tris Base, High Purity, Low Endotoxin, Low Metals, USP, EP, BP, ChP (T-153) 1 kg GMP, Multicompandial CAS: 77-86-1
Supplier: Pfanstiehl Inc T1531KG
Tris Base iscommonly used buffer in biologics production and various biochemical and molecular biology applications. It is a zwitterionic amine, meaning it has both acidic and basic functional groups in its structure, making it an effective buffer over a wide pH range.