Learn More
Electron Microscopy Sciences Tri-Pour Beaker Cap for 250 ml beaker
SDP
Supplier:  Electron Microscopy Sciences 60973
Encompass_Preferred
The caps are available for all Tri-Pour beakers. The caps are wax impregnated, have lift-tabs, and may be written on for sample identification.
Use: 250 ml
Catalog No. 50-996-327
Explore available promotions