Learn More
XpressBio TRANSIL Microsomal Binding Kit
SDP
Supplier:  XpressBio TMP01202096
Encompass_Preferred
The TRANSIL Microsomal Binding kit measures the affinity of drugs to human microsomal membranes and determines microsomal binding in stability incubation experiments. This allows the accurate estimation of intrinsic clearance from stability incubations by correcting the experimental clearance with the fraction of drug unbound in the incubation. Internal quality controls provide easy assessment of recovery, experiment and data quality.
Catalog No. NC2333881
Explore available promotions