Learn More
Perkin Elmer US LLC Scott/Cross-Flow Sample Introduction Torch Module for Optima 2x00/4x00/5x00/7x00 DV
Supplier: Perkin Elmer US LLC N0770606
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This module kit includes: Inner Chamber for External Ryton/Type II Spray Chamber, Qty. 1 (N0582258) Outer Axial Spray Chamber, Qty. 1 (N0775296) 1-Slot Quartz Torch, Qty. 1 (N0770338) GemTip Cross-Flow II Nebulizer, Qty. 1 (N0770546) 2.0 mm I.D. Alumina Injector (N0775177) Quick-Change Torch Module Mounting Block, Qty. 1 (N0770600) Scott Spray Chamber External Drain Kit, Qty. 1 (N0770613)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.