Learn More
Hach Company KF1000 Volumetric Titrator for Karl Fischer Titrations
Supplier:  Hach Company KF1121.97
Encompass_Preferred
Request Bulk Order
The KF1000 is an accurate, reliable and easy to use instrument for performing volumetric titrations in the laboratory. This titrator was designed in Germany as part of the Hach Titralab family of automatic titrators and allows you to quickly measure moisture content. Moisture can affect product quality, texture, shelf life, chemical stability and reactivity, and the Karl Fischer volumetric titration is a universally acknowledged method used in all types of substances (including chemicals, oils, pharmaceuticals and food). If you perform volumetric titrations to determine moisture content this is the only instrument you need to consider! The KF1000 instrument was designed to be used with Hach’s Karl Fischer Moisture Content Application Pack (Hach p/n AP0014.KF1121) which needs to be purchased separately. The application pack includes the pre-programmed application that runs on the instrument as well as a set of accessories necessary to perform your volumetric titrations.
Catalog No. 50-199-7736
Due to product restrictions, please to purchase or view availability for this product.
Explore available promotions