Learn More
TCI America™ Tissue-Clearing Reagent CUBIC-HL
SDP
CUBIC-HL is optimized for clearing highly fatty tissues. Note this does not preserve the fluorescent signal of fluorescent proteins.
Supplier:  TCI America™ T3781100ML
 View more versions of this product
Catalog No. T3781100ML
Add to Cart
Description
Description
  • The CUBIC technique enables cyclopedic imaging at a single-cell resolution following whole-body and whole-organ clearing.
  • CUBIC-HL is an optional addition to the CUBIC clearing protocol, and can be used to clear highly fatty tissue and quenching autofluourescence.
Note: Both T3741 and T3740 are required for the CUBIC clearing process.
Specifications
Specifications
100 mL
CUBIC
Liquid
Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device
Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications
Product Suggestions
Product Suggestions