Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Watson Gloves Thrasher Welding Gloves
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- Split cowhide leather
- Stitched with Aramid Fiber thread for strength and durability
- Cutshield™ P-aramid/steel/polyester liner
- Full leather index finger and inset thumb
- Gauntlet style cuff
- Snug-fitting shirred elastic wrist
- ANSI Level 2 rating can withstand up to 140°C/284°F for at least 15 seconds.
Specifications
Back Material | Split Cow Hide Leather |
Certifications/Compliance | ANSI Cut, ANSI Heat, ANSI Puncture |
Chemotherapeutic Rated | No |
Cuff Material | Leather Gore |
Color | Brown, Black |
Grade | Premium |
Hand | Ambidextrous |
Material | Split Cowhide Leather |
Product Type | Welding Gloves |
Size | X-Large |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction