Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
The TruScan RM handheld Raman spectrometer and the microPHAZIR RX handheld NIR material analyzer provide reliable results within seocnds.
TruScan RM with Discovery Library (Mfr. No. 800-00391-01)
- Features highly selective Raman spectroscopy
- Fast, point of need raw material identification (10-15 seconds)
- Portable handheld and weighs less than 2 lbs (0.9kg)
- Clear PASS or FAIL results enable user independence and require no secondary interpretation
- Broad compound coverage includes cellulose based products
- Fast and easy method development through assisted signature/reference acquisition
- Intuitive workflow adapted to production environment through batch reports and ease of data input using barcode scanner
- Greatly reduces analysis cost and overall overhead
- Enhanced 21 CFR Part 11 and GMP compliance with biometric login, complex password options, and full audit trail features
- Non-contact analysis through plastic bags, glass containers, blister packs, and clear gel caps greatly eliminates potential staff exposure to hazardous chemicals and highly potent active pharmaceutical ingredients (API) during inspection
- Easy fleet management feature enable instrument and identification methods cloning
- 24/7/365 Live customer support
- Battery operated instrument (4 hours on a single charge)
- Qualification documentation and standard operating procedures available
- Portable handheld solution
- Lightweight 2.75 lb (1.25kg) device
- Intuitive Color LCD screen
- Rechargeable Li-Ion batteries
- Long battery operation
- Simple operation, no experience required
- Minimal training required
- Instantaneous (1-2 seconds) results
- Nondestructive measurement
- Results are logged in a on-board database
- Fully programmable via PC software
- Ready-to-use library with 100s of ingredients
- Integrated barcode scanner
- 21 CFR Part 11 compliant
- IQ/OQ/PQ qualification package
- USP 1120 and EP 2.2.40 compliant
- EMEA & PIC/S Compliant
- Traceable standards available
- Validation services available
Specifications
TruScan RM and micrPHAZIR RX Package | |
Rechargeable Lithium Ion Battery | |
Barcode Supported Symbologies, Biometrics | |
0.9 kg |
LCD | |
21 CFR Part 11 and GMP, USP 1120 and EP 2.2.40, EMEA and PIC/S | |
2 lb. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction