Learn More
Thermo Scientific™ RadEye™ HEC Alpha/Beta Sample Counter
Stand-alone scaler counter
Supplier:  Thermo Scientific™ 4250697
Request Bulk Order
Catalog No. 19-155-222
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Explore available promotions
Description
Description
The RadEye HEC is a sample counting system that provides simultaneous alpha and beta measurements.
Features
  • Simultaneous alpha/beta measurements
  • 800 hours battery operation
  • Non-volatile data storage
  • Customized library of up to 16 test sources with automated half-life correction
  • Library of up to 16 nuclide efficiencies
  • Simple detector performance verification with 9g Lutetium Test Adapter
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions