Learn More
Thermo Scientific™ Transport Case for PackEye Radiation Detection Backpack
Supplier: Thermo Scientific™ 4255085
Description
- Allows the inside operation of a PackEye to be used as stationary detector
- Case-adapter provides external power for PackEye
- Use the optional PackEye PDA for data display
- PackEye not included
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Connector for external power supply / charging |