Learn More
Thermo Scientific™ MAS™ LabLink xL COMMUNICATOR and CLOUD COMMUNICATOR
Facilitates the automatic upload of QC data from various clinical laboratory instruments, middlewares and laboratory information systems to LabLink xL WEB
Supplier: Thermo Scientific™ LLXLF
This item is not returnable.
View return policy
Description
- Fully automated, real time QC data transfer via Data Innovations Instrument Manager and Beckman REMISOL Advance Data Manager to LabLink xL
- Numerous semi-automated connectivity options available for QC data upload from instrument, LIS or middleware to LabLink xL
- Hassle-free setup
- Simple and easy to use
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)