Learn More
Aat Bioquest Texas Red-DHPE CAS 187099-99-6 (1 mg)
Supplier: Aat Bioquest 23300
Texas Red DHPE is a phospholipid that is labeled on the head group with the bright red-fluorescent Texas Red dye excitation/emission maxima 595/615 nm. This catalog is for 1 mg.
This item is not returnable.
View return policy