Learn More
Quantimetrix QuantTest Red Pyrogallol Red Reagent
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Quantimetrix 521012
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
• For quantitative determination of protein in cerebrospinal fluid and urine for both manual and automated systems • Reagents stable until the expiration date when stored properly • Pyrogallol Red Molybdate dye binding method provides a simple, ready-to-use colorimetric method for total protein quantitation with greater linearity, using microliter samples of urine or cerebrospinal fluid (CSF) in manual or automated systems.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.