Learn More
VHG Labs SULFUR IN MINERAL OIL 25
Supplier:  VHG Labs VHGSMIN25100
Encompass_Preferred
VHG-SMIN-25-100- Sulfur Standard: S @ 25 µg/g (0.0025 wt%) in 75 cSt Mineral Oil
Catalog No. NC1658957
Explore available promotions