Learn More
Showa Denko America Inc SUGAR SP0810 8.0X300MM, 7MICRN
Supplier: Showa Denko America Inc F6378105
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
These columns separates saccharides by the comination of ligand exchange and size exclusion modes. Counterion: Pb(2+).