Learn More
Showa Denko America Inc SUGAR KS-801
Supplier: Showa Denko America Inc F6378010
Styrene divinylbenzene based column separates saccharides by combination of ligand exchange and size exclusion modes, contains sodium counterion, fulfills USP L22 and L58 requirements, can be used in combination with other KS columns, only water is required for analysis of neutral sugars, dimensions: 8.0 mm ID x 300 mm L