Learn More
Streck Sickledex™ Solubility Kit
![SDP](/content/dam/fishersci/glyphs/sdp.png)
SICKLEDEX is a qualitative solubility test that can detect the presence of sickling hemoglobin in human blood samples within 6 minutes. With this kit, your laboratory is supplied with items necessary to perform easy and efficient sickle solubility testing, including the SICKLEDEX solubility test kit, Sickle-Chex™ whole blood control and a convenient test tube rack.
$202.00 - $501.00
Specifications
Type | Sickle Cell Solubility Test Kit |
---|---|
Includes | 2 Bottles of Phosphate Buffer with Saponin, 2 Buffer Dispensing Caps, 2 Vials of Sodium Hydrosulfite Reagent Powder |
Test Time | 6 min. |
Includes: Each test kit contains two bottles of phosphate buffer with Saponin, two buffer dispensing caps and two vials of Sodium Hydrosulfite reagent powder.
Requires: Each test requires 2 mL of reconstituted solubility buffer and 20μL of anticoagulated whole blood or 20μL of sickle cell hemoglobin control.
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-285-555
![]() |
Streck
217661 |
12 Tests |
Pack of 12 for $202.00
|
|
|||||
23-285-625
![]() |
Streck
217660 |
50 Tests |
Pack of 50 for $386.00
|
|
|||||
11-716-450
![]() |
Streck
217657 |
100 Tests |
Pack of 100 for $501.00
|
|
|||||
Description
- Test results available in 6 minutes.
- Requires only 20 µL of patient or control sample.
- 45-day open-vial stability, 12-month closed-vial stability.
- Buffer packaging reduces waste and maximizes open-vial stability.
- No pipetting necessary.
- Sickle-Chex whole blood control can be used like a patient sample.
Order Info
Additional test tube racks are available separately, Cat. No. 11-716-452. Additional test tubes, Cat. No. NC1459622.
Specifications
Sickle Cell Solubility Test Kit | |
2 Bottles of Phosphate Buffer with Saponin, 2 Buffer Dispensing Caps, 2 Vials of Sodium Hydrosulfite Reagent Powder | |
6 min. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.