Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streamlight™ Rechargeable E-Spot™ LiteBox™ Lanterns
![SDP](/content/dam/fishersci/glyphs/sdp.png)
The E-Spot Litebox is an industrial-duty, rechargeable lantern that features a bright 540 lumens of light in a spot beam pattern that reaches 469 meters.
Supplier: Streamlight™ 45851
This item is not returnable.
View return policy
Description
- 540 lumens on high
- 469 meter beam distance
- Choose low mode for a run time of 15 hours
- Swivel neck allows you to aim the beam where you need it
- IPX4 rated for weather resistant operation
- Jumper-selectable to high or low setting
Specifications
Lantern | |
50,000 hr. | |
3583.38 g | |
Lead Acid | |
Orange | |
540 lm |
3 x C4 LED | |
7 lb. 14.40 oz. | |
7 hr. (high), 15 hr. (low) | |
469 m | |
Strap, AC/DC power chargers, and mounting rack | |
ABS Thermoplastic |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction