Learn More
Pelican Products Inc. STORM CASE W/FOAM BLACK IM2075
Supplier:  Pelican Products Inc. IM207500001
Encompass_Preferred
DIMENSIONSInterior (L×W×D)9.50 x 7.50 x 7.25 in (24.1 x 19.1 x 18.4 cm)Exterior (L×W×D)11.80 x 9.80 x 7.70 in (30 x 24.9 x 19.6 cm)MEASUREMENTSLid Depth1.50" (3.8 cm)Bottom Depth5.75" (14.6 cm)Total Depth7.25" (18.4 cm)Int Volume0.3 ft³ (0.008 m³)Padlock Hole Diameter0.28" (0.7 cm)WEIGHTSWeight With Foam3.70 lbs (1.7 kg)Weight Empty3.30 lbs (1.5 kg)Buoyancy18.20 lbs (8.3 kg)MATERIALSBody MaterialInjection Molded HPX™ high performance resinLatch MaterialValox - Polybutylene Terephthalate (PBT)Latch (Alt) MaterialXenoy - Polyester/Polycarbonate blendO-Ring MaterialEPDMPins MaterialStainless SteelPins (Alt) MaterialAluminumFoam Material1.3/1.35 polyesterPurge Body MaterialValox - Polybutylene Terephthalate (PBT)Purge Body (Alt) MaterialXenoy - Polyester/Polycarbonate blendPurge Vent MaterialGortex membraneTEMPERATURESMinimum Temperature-20° F (-29 ° C)Maximum Temperature140° F (60 ° C)APPROVALSCertificationsIP67
Catalog No. NC2888088
Explore available promotions