Learn More
Sterlitech Corporation Kit For Kato-Katz Method, 500 Tests/Box
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Sterlitech Corporation 2050020
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Contents of Kit: Applicator Sticks (500 pieces) Nylon Screen, 100 mesh (500 pieces) Template, calibrated to 41.7mg (500 pieces) Hydrophilic Cellophane (500 pieces) Additional Items needed: Microscope Slides (500) Flat-bottomed jar Forceps Absorbent Tissue Newspaper Glycerol/Dye Solution (1 mL 3% Malachite Green or Methylene Blue in 200 mL of >50% glycerol solution)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.