Learn More
Sklar Instruments Econo(TM) Sterile Iris Forceps, Sklar , 4 in., Half Curved, Serrated, Stainless Steel, Disposable, Sterile. CS/25.
SDP
Supplier:  Sklar Instruments 962515
Encompass_Preferred
Econo(TM) Sterile Iris Forceps, Sklar, 4 in., Half Curved, Serrated, Stainless Steel, Disposable, Sterile. Econo Sterile(TM) Iris Dressing Forceps are thumb style forceps commonly used for placing or removing dressings in many types of procedures. Although Dressing Forceps were originally designed to specifically hold materials such as gauze, they have come to have a variety of additional functions. They may be used for removing foreign objects or dead tissue from wounds, as well as other small surgical materials such as tubing. Serrated jaws assist in a having a secure hold on items. Econo(TM) Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments made in Pakistan. Econo(TM) Sterile instruments eliminate the need for cleaning and reduce the likelihood of cross-contamination. BX25.
Catalog No. NC0691714
Explore available promotions