Learn More
Instech STER LOCK SOLN HEP 50ML W/PP
SDP
Supplier:  Instech USPHDS50050VYP1
Encompass_Preferred
Pharmaceutical-grade catheter lock solution prepared with heparin sodium following the USP <797> standard for compounding sterile preparations. The solution contains heparin (500 USP units/ml) in 50% dextrose, one 50ml vial, yellow, with PinPort™ access.
Catalog No. NC2816901