Learn More
Tri Continent Scientific StatMatic I, 8-Channel Microplate Washer And Bottle Top Dispenser
Supplier:  Tri Continent Scientific 0982
Encompass_Preferred
The Next Best Thing to Automation. The StatMatic I microplate washer and bottle top dispenser provides a convenient and affordable alternative for diagnostic assays and cell culture work. Used with an eight-port disposable manifold, the StatMatic I microplate washer and bottle top dispenser provides continuous operation not possible with multi-channel pipettes. Eight-port manifold, designed for 96-well microplates, delivers fluids in volumes of 100, 200, 250, or 300 L with well-to-well accuracy of 5% or better; Constructed of polypropylene, Teflon, glass, and ceramics, there is no exposed glass that can break and requires minimal maintenance. ; Compatible with most reagents and chemicals and designed so that even protein-based reagents won't cause sticking. ; Filtered air vent keeps liquids free of contaminants. ; Priming line allows recirculation into the dispenser port without removing the bottle.
Catalog No. NC0654011
Explore available promotions