Learn More
EKF Diagnostics STAT-Site™ Blood Glucose Strip, 25 Count
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Glusose strip that provides result in 5 seconds
Supplier: EKF Diagnostics 7010025
This item is not returnable.
View return policy
Description
- 1 μL sample size
- Uses capillary or venous whole blood
Specifications
Test Strip |