Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stanbio™ LiquiColor™ Albumin Test
For Bromocresol Green reaction
Supplier: Stanbio™ 0285500
Includes: Standard: 1 x 3mL (6g/dL), reagent: 2 x 250mL
Description
- Ready-to-use liquid reagent
- Linear to 7g/dL
- Read at 550nm
- Volume: 500mL
Specifications
LiquiColor™ Albumin Test | |
Ready-to-use liquid reagent | |
1 x 3mL Standard, 2 x 250mL reagent | |
500 mL |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction