Learn More
Wilmad Labglass STALAGMOMETER TUBE ONLY 3.5ML
Supplier: Wilmad Labglass LG5050104
Stalagmometer, Tube Only. Size: 3.5mL. Specifically designed for checking the surface tension of liquids by drop count method.
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov