Learn More
Seca Corp 535 Spot Check Vital Signs Monitor (NiBP, Oral Temp)
SDP
Supplier:  Seca Corp 5351010131
Encompass_Preferred
The Spot-check-monitor to precisely measure the primary three vital signs: blood pressure, pulse rate, and oral temperature. Measurement results on the touchscreen monitor. Large internal memory for up to 70,000 measurements. • Essential vital signs measurement of blood pressure, pulse and oral temperature • Easy-to-understand graphical presentation of the measurement results on the touchscreen monitor • Large internal memory for up to 70,000 measurements • EMR-validated to any EMR system • Weight: 5.9 lbs • Dimensions (WxHxD) 9.9 x 10.3 x 10.9 inches • Display: 7-inch touchscreen • Connectivity: Wi-Fi, Ethernet • Ports: 2 x USB 2.0 • Software: seca Connect 103 • Component options • Temperature probe (oral/axillary) • Accessories • Rolling stand seca 475 • Carrying case seca 432
Catalog No. NC1834740
Explore available promotions