Learn More
AVANTOR MASTERFLEX (VWR LLC) John Guest PPSV010822W Speedfit Ball Valve, straight, 1/4"" OD speedfit x 1/4"" NPT(M)
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX0739100
Food-grade polypropylene housing is specifically made for use with potable water. Quickly disconnect and reconnect your water lines with these Speedfit valves.