Learn More
AVANTOR MASTERFLEX (VWR LLC) John Guest PPSV500822W Speedfit Ball Valve, elbow, 1/4"" OD speedfit x 1/4"" NPT(F)
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX0739108
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Food-grade polypropylene housing is specifically made for use with potable water. Quickly disconnect and reconnect your water lines with these Speedfit valves.