Learn More
CTL Scientific Supply Corp SPEC IND PH 1-14,PK/6 RL X16
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: CTL Scientific Supply Corp 902046
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Universal pH paper 1-14, pack of 6 rolls (each 5 meters length) in dispensers with color charts. Gradation of scale: 1 - 2 - 3 - 5 - 6 - 7 - 8 - 9 - 10 - 12 - 14 for extra rolls see Refill 90224