Learn More
CTL Scientific Supply Corp SP IND PH 7.2-9.7,PK/6 RL X16
Supplier: CTL Scientific Supply Corp 902116
Special Indicator pH paper 7.2-9.7, pack of 6 rolls (each 5 meters length) in dispensers with color charts. Gradation of scale: <7.2 - 7.2 - 7.5 - 7.8 - 8.1 - 8.4 - 8.7 - 9.0 - 9.3 - 9.7 - >9.7