Learn More
Supertek™ Solar Powered Car
Explore Green Energy on a small scale with this Solar Powered Car.
Supplier: Supertek™ PH50608S3
Description
- Operates on sunlight or a rechargeable AA battery.
- A four-position switch allows you to choose between running the car on the solar cell or the battery - recharging the battery from the solar cell - or off.
- Two binding posts allow the voltage developed by the solar cell to be measured in all four switch positions.
- The solar cell can be titled to show the effect of the suns angle on the solar cell voltage.
- The car can also be operated using a strong lamp.
- Approx. 6 in. x 2.75 in. x 3 in.
- Includes rechargeable battery.
- Ages 7 plus
Specifications
Solar Car |