Learn More
Clontech Labs 3P SMARTScribe™ Reverse Transcriptase, 40 Rxns
Supplier: Clontech Labs 3P 639536
SMARTScribe Reverse Transcriptase is a modified Moloney Murine Leukemia Virus Reverse Transcriptase that synthesizes full-length cDNA from rare or long transcripts. SMARTScribe RT preparations are exceptionally pure, with all contaminating nucleases removed. The enzyme is useful for a wide range of applications, and has been specially formulated for use with all of our SMART Kits. 40 Rxns
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.