Learn More
Thermo Scientific™ Smart-Vue Pro Modules
Increase efficiency with multi-sensor wireless monitoring
Supplier: Thermo Scientific™ SVPHWRMOD014
Description
Smart-Vue Pro modules are available in two configurations- Duo and Quatro. The Duo allows up to 2 sensor channels for monitoring. The Quatro has extended range of 4 sensor channels for 2 wired and 2 wireless sensors. Ideal for monitoring several pieces of equipment simultaneously or multiple points within one piece of equipment
- Bright 2.4 in. color touch screen with multiple views
- Alarms locally with real -time audio-visual alarms on the modules
- Audio visual alarms for local notification
- Communication locally via Bluetooth and to the system via LoRaLow
- AC power consumption through USB port access
- Optional replaceable battery power with extended life (minimum of 1 year) allows continuous monitoring
- Choice of wired and wireless sensors with automatic sensor detection
- User traceability with customized levels of user access with encrypted password protection
Specifications
Module | |
3.6 V lithium battery | |
Two 3.6 V lithium batteries, two sensor mounting kits, one mounting holder, one antenna, and two 1.5 m/5 ft. flat cables |
LCD | |
2 years | |
Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance |