Learn More
Thermo Scientific™ Smart-Vue Pro Modules
Increase efficiency with multi-sensor wireless monitoring
Supplier:  Thermo Scientific™ SVPHWRMOD012
 View more versions of this product
Catalog No. SVPHWRMOD12
Explore available promotions
Add to Cart
Description
Description
Smart-Vue Pro modules are available in two configurations- Duo and Quatro. The Duo allows up to 2 sensor channels for monitoring. The Quatro has extended range of 4 sensor channels for 2 wired and 2 wireless sensors. Ideal for monitoring several pieces of equipment simultaneously or multiple points within one piece of equipment
  • Bright 2.4 in. color touch screen with multiple views
  • Alarms locally with real -time audio-visual alarms on the modules
  • Audio visual alarms for local notification
  • Communication locally via Bluetooth and to the system via LoRaLow
  • AC power consumption through USB port access
  • Optional replaceable battery power with extended life (minimum of 1 year) allows continuous monitoring
  • Choice of wired and wireless sensors with automatic sensor detection
  • User traceability with customized levels of user access with encrypted password protection
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Module
3.6 V Lithium Battery
Two 3.6 V Lithium batteries, two sensor mounting kits, one mounting holder, one antenna, and two 1.5 m/5 ft. flat cables
LCD
2 Years
Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Product Suggestions
Product Suggestions