Learn More
Greiner Bio-One Small Volume™ 384 Deep Well Polypropylene Microplates
Ensures accurate pipetting, reduces dead volume. Greiner Bio-One Small Volume™ 384 Deep Well Polypropylene Microplates are specifically designed to accommodate heat-sealing systems.
Supplier: Greiner Bio-One 784209
Description
- Non-pyrogenic
- Conical well design with flat bottom
- Ensures accurate pipetting
- Reduces dead volume of well
- Square well geometry, compatible with CapMat plate sealers
- Accommodates heat-sealing systems
-
Service Plan
No
Warranty and Services
No
Specifications
Standard | |
Black | |
384 | |
1 to 90 μL |
16 x 24 | |
Polypropylene | |
Non-sterile | |
Deep well |