Learn More
Cayman Chemical SM-102, CC-33474-100
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Cayman Chemical 33474100
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
SM-102 is a cationic lipid used for transfection and formulation of nucleic acid-based therapeutics. It is supplied as a solution in ethanol at 10 mg/mL concentration by Bionordika. Some key features of SM-102 include its ability to form small and stable nucleic acid-containing nanoparticles, enable effective cytosolic delivery and endosomal escape of nucleic acids, and exhibit low toxicity. SM-102 is compatible with various nucleic acid payloads including mRNA, siRNA, and DNA. The 100 mg quantity is sufficient for formulating 10 mL of 10 mg/mL liposomes. SM-102 can facilitate effective delivery of nucleic acid therapeutics in vitro and in vivo.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.