Learn More
Chemglass Life Sciences Sleeve, Clamp Replacement, Medium Two-Prong, Vinyl
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CG923901
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Sleeve, Clamp Replacement, Medium Two-Prong, Vinyl. Replacement sleeves for two and three prong clamps. Made of vinyl or fiberglass.