Learn More
Sklar Instruments Econo Sterile Dressing Forceps - Box of 25 - 8"
Supplier: Sklar Instruments 962921
Length: 8" | Tip Configuration: Blunt | Instrument Type: Dressing Forceps | Curvature: Straight | Working Surface Style: Serrated | Handle: Thumb | Material: Stainless Steel | Disposable | Sterile
This item is not returnable.
View return policy