Learn More
Thermo Scientific 4.5 mm (Robust) Skimmer Cone Insert
Supplier: Thermo Scientific BRE0006591
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
3.5 mm Skimmer Cone insert for iCAP Q/Qnova ICP-MS Systems – Robust Matrix Tolerance Applications
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)