Learn More
CTL Scientific Supply Corp SILVER TEST PAPER,200 STRIPS
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: CTL Scientific Supply Corp 90732
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
SILVER TEST PAPER - 200 test strips. Test paper shows silver ions (Ag+) as rot-violet spots on a salmon-red background. Interferences caused by mercury and copper ions are readily circumvented. Limit of sensitivity:20 mg/l Ag +