Learn More
W.S. TYLER™ Sieve Certification and Recertification Services
Certification services offered for new or used sieves
$212.00
Specifications
For Use With (Application) To serialize and supply with documentation
View More Specs
Products 1
Catalog Number Mfr. No. dimProductType Price Quantity & Availability  
Catalog Number Mfr. No. dimProductType Price Quantity & Availability  
04-909-16
View Documents
W.S. TYLER™
9998
Each of 1 for $212.00
Add to Cart
Description
Description
Certification Service
  • Order when purchasing new sieves
  • Uses a computerized video imaging system to verify that each sieve opening meets ASTM™ and ISO specifications within a tolerance of ±0.1 microns
  • Sieves are then serialized and supplied with documentation, traceable to NIST™, detailing the openings and the wire diameters of the mesh
Recertification Services
  • Offer the same examination and documentation processes for in-use sieves as for new sieves
  • Contact your Fisher Scientific Customer Service Representative for the recommended sieve aperture/recertification schedule
Specifications
Specifications
To serialize and supply with documentation