Learn More
Hach Company Safety Shield
Supplier: Hach Company 5004000
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For use with Hach Digesdhal Apparatus or COD Reactor. Order 5003000 for COD Reactor Polycarbonate Shield 15"Height x 16" Width. Order 5004000 for Hach Digesdhal Polycarbonate Shield 30" Height x 16" Width. Heavy 2.223 cm thick (0.875") urethane coated base resists slipping on countertops and lab benches. Shield formed in a parabolic curve.