Learn More
Ace Glass, Inc. Mini-Lab™; short path still, 10mL, complete with distillation flask, gas inlet tube adapter, 14/20 still body, cow receiver and (3)5mL pear-shaped receiving flasks
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ace Glass, Inc. 931620
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
10mL Mini-Lab™ short path distillation apparatus. Sold complete with distillation flask, gas inlet tube adapter, 14/20 still body, cow receiver, and (3) 5mL pear-shaped receiving flasks. Apparatus is designed for small quantities where holdup between flask and receiver must be minimized but where it is also desired to measure vapor temperature with an accuracy compared to large set-ups. Apparatus has improved thermometer sensitivity. Physical carry-over is practically eliminated by impingement. Condenser delivery tube is straight through. Condensate drops directly into one of the four receivers attached to cow. Upper hose connections are for water lines, lower connection is for vacuum/vent. Apparatus has a 10/30 thermometer joint, and all other joints are 14/20.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.