Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Visby Medical™ Sexual Health Collection Kit
Vaginal specimen collection kit for use with the Sexual Health Test by Visby Medical™. Kit contains 50 swabs, 50 tubes containing collection media, and 50 self-collection instructions.
Supplier: Visby Medical™ PS000715
Description
- For use with the Visby Medical™ Sexual Health Test (PS-400372).
- Contains 50 vaginal specimen self-collection kits with swabs/tubes/self-collection instructions.
- Designed to collect, preserve, and transport Chlamydia trachomatis (CT), Neisseria gonorrhoeae (NG), and Trichomonas vaginalis (TV) in patient-collected vaginal specimens from symptomatic and asymptomatic women prior to analysis with the Visby Medical™ Sexual Health Test.
Specifications
Non-sterile |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction